Tisk…

methylcyklohexan 99%, Spectrophotometric Grade, Riedel-de Haën™

Dodavatel: Honeywell Chemicals
Nebezpečí

259691-1L 259691-2L
259691-1LEA 7940 CZK
259691-1L 259691-2L
methylcyklohexan 99%, Spectrophotometric Grade, Riedel-de Haën™
methylcyklohexan

  • Spectrophotometric grade solvents are high purity solvents made specifically for applications using spectrophotometers.


Beilstein Number: 505972
RTECS Number: GV6125000
UNSPSC Code: 12190000

Vzorec: C₆H₁₁CH₃
Bod varu: 100…102 °C (1,013 hPa)
Bod tání: –126 °C
Hustota: 0,769 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001497
CAS číslo: 108-87-2
EINECS: 203-624-3
UN: 2296
ADR: 3,II
Merck Index: 13,06074
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f7f0d5a1c32138da16df4781897a9c0184e42b7&vid=39a9392c8629d0e2fc79134840e6d8a8dc8a76e7&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Identity (IR) complying
Assay (GC) Min. 98.5 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Absorbance at 400 nm Max. 0.01
Absorbance at 300 nm Max. 0.01
Absorbance at 260 nm Max. 0.05
Absorbance at 232 nm Max. 0.15
Absorbance at 221 nm Max. 0.40
Absorbance at 207 nm Max. 1.0
Explosion Limit 6.7 %
Vapor Pressure 83.2 mmHg ( 37.7 °C)

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR