Ethylcyclohexane, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
E19154-100G
E19154-25G
SIALE19154-100GEA
5650
CZK
SIALE19154-100G
SIALE19154-25G
Ethylcyclohexane, Sigma-Aldrich®
Ethylcyclohexane
Vzorec:
C₈H₁₆ Bod varu: 132 °C (1013 hPa) Hustota: 0,788 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001523 CAS číslo: 1678-91-7 EINECS: 216-835-0 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...