Tisk…

1-Cyclohexylethanol ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B20363.06 B20363.14
B20363.06EA 3400 CZK
B20363.06 B20363.14
1-Cyclohexylethanol ≥98%
1-Cyclohexylethanol

Purity: 98% CAS No.: 1193-81-3 Molecular Formula: C8H16O Formula Weight: 128.22 Flash Point: 73°(163°F) Refractive Index: 1.4650 Boiling Point: 188-190° MDL No.: MFCD00001475

Vzorec: C₈H₁₆O
MW: 128.21
Bod varu: 188…190 °C
Hustota: 0.918
Bod vzplanutí: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001475
CAS číslo: 1193-81-3
EINECS: 214-780-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87b02ed7f0ec80c1d02a0b5414c0585a6d8fadd9&vid=34a65012f79e7eb749c7cecc0c01e3a2172463a3&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR