Tisk…

trans-4-Methylcyclohexanecarboxylic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: trans-4-Methyl-1-cyclohexanecarboxylic acid

M-Clarity™ quality level = MQ100

331600-1G 331600-5G
SIAL331600-1GEA 6510 CZK
SIAL331600-1G SIAL331600-5G
trans-4-Methylcyclohexanecarboxylic acid, Sigma-Aldrich®
trans-4-Methylcyclohexanecarboxylic acid
Vzorec: C₈H₁₄O₂
MW: 142,2 g/mol
MDL Number: MFCD00074943
CAS číslo: 13064-83-0
EINECS: 235-959-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR