Tisk…

3,4-Dimethyl-1,2-cyclopentanedione, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3,4-Dimethylcyclopentane-1,2-dione, 3,4-Dimethyl-1,2-cyclopentadione

M-Clarity™ quality level = MQ300

W326801-100G-K W326801-1KG-K W326801-25G-K
SIALW326801-100G-KEA 15060 CZK
SIALW326801-100G-K SIALW326801-1KG-K SIALW326801-25G-K
3,4-Dimethyl-1,2-cyclopentanedione, Sigma-Aldrich®
3,4-Dimethyl-1,2-cyclopentanedione
Vzorec: C₇H₁₀O₂
MW: 126,16 g/mol
Bod varu: 219 °C (1013 hPa)
Bod tání: 71…72 °C
MDL Number: MFCD00143063
CAS číslo: 13494-06-9
EINECS: 236-810-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR