(Chloromethyl)cyclopropane, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Cyclopropyl methyl chloride
M-Clarity™ quality level = MQ100
184667-10G
184667-50G
SIAL184667-10GEA
8960
CZK
SIAL184667-10G
SIAL184667-50G
(Chloromethyl)cyclopropane, Sigma-Aldrich®
(Chloromethyl)cyclopropane
Vzorec:
C₄H₇Cl MW: 90,55 g/mol Bod varu: 87 °C (1013 hPa) Bod tání: –91 °C Hustota: 0,98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001307 CAS číslo: 5911-08-0 EINECS: 227-632-1 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...