(Bromomethyl)cyclopropane, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
242403-1G
242403-5G
SIAL242403-1GEA
1440
CZK
SIAL242403-1G
SIAL242403-5G
(Bromomethyl)cyclopropane, Sigma-Aldrich®
(Bromomethyl)cyclopropane
Vzorec:
C₄H₇Br Bod varu: 105 °C (1013 hPa) Hustota: 1,41 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001306 CAS číslo: 7051-34-5 EINECS: 230-331-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...