(Bromomethyl)cyclohexane, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
C106003-100G
C106003-25G
C106003-5G
SIALC106003-100GEA
7440
CZK
SIALC106003-100G
SIALC106003-25G
SIALC106003-5G
(Bromomethyl)cyclohexane, Sigma-Aldrich®
(Bromomethyl)cyclohexane
Vzorec:
C₇H₁₃Br Bod varu: 100 °C (53,3 hPa) Hustota: 1,269 g/cm³ (20 °C) Bod vzplanutí: 57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001509 CAS číslo: 2550-36-9 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...