Tisk…

Ethynylcyklopropan, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Cyclopropylacetylene

MQ100

663018-25G 663018-5G
SIAL663018-25GEA 3540 CZK
SIAL663018-25G SIAL663018-5G
Ethynylcyklopropan, Sigma-Aldrich®
Ethynylcyklopropan
Vzorec: C₅H₆
MW: 66.1
MDL Number: MFCD02181090
CAS číslo: 6746-94-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR