1-Ethynylcyclohexanol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1-Ethynyl-1-cyclohexanol
M-Clarity™ quality level = MQ200
E51406-100ML
E51406-20L
E51406-5L
SIALE51406-100MLEA
1460
CZK
SIALE51406-100ML
SIALE51406-20L
SIALE51406-5L
1-Ethynylcyclohexanol, Sigma-Aldrich®
1-Ethynylcyclohexanol
Vzorec:
C₈H₁₂O Bod varu: 180 °C (1013 hPa) Bod tání: 30…32 °C Hustota: 0,973 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003858 CAS číslo: 78-27-3 EINECS: 201-100-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...