Cycloheptane, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
C98403-25G
C98403-5G
SIALC98403-25GEA
0
CZK
SIALC98403-25G
SIALC98403-5G
Cycloheptane, Sigma-Aldrich®
Cycloheptane
Vzorec:
C₇H₁₄ MW: 98,19 g/mol Bod varu: 120 °C (1013 hPa) Bod tání: –12 °C Hustota: 0,8093 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004147 CAS číslo: 291-64-5 EINECS: 206-030-2 UN: 1206 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...