Tisk…

myo-Inozitol, Millipore®

Dodavatel: Merck

Synonyms: i-Inositol, D-Myo-Inositol, rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaol, Inositol

M-Clarity™ quality level = MQ200

57570-100G 57570-25G 57570-500G
SIAL57570-100GEA 0 CZK
SIAL57570-100G SIAL57570-25G SIAL57570-500G
myo-Inozitol, Millipore®
myo-Inozitol
Vzorec: C₆H₁₂O₆
MW: 180,16 g/mol
Bod varu: 291 °C (1013 hPa)
Bod tání: 210 °C
Hustota: 1,752 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00077932
CAS číslo: 87-89-8
EINECS: 201-781-2
Merck Index: 13,05001

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR