myo-Inozitol ≥98%
Dodavatel: Thermo Scientific
Synonyms:
i-Inositol, D-Myo-Inositol, rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaol, Inositol
A13586.0E
A13586.22
A13586.36
A13586.0EEA
18120
CZK
A13586.0E
A13586.22
A13586.36
myo-Inozitol ≥98%
myo-Inozitol
Purity: 98+%
CAS No.: 87-89-8
Molecular Formula: C6H12O6
Formula Weight: 180.16
Physical Form: Cryst powder
Density: 1.75
Appearance: White
Odour: Odourless
Melting Point: 224-227°
Beilstein Reference: 1907329
Merck Reference: 14,4978
MDL No.: MFCD00077932
A lipotropic agent
Vzorec:
C₆H₁₂O₆ MW: 180,16 g/mol Bod varu: 291 °C (1013 hPa) Bod tání: 224…227 °C Hustota: 1,75 Storage Temperature: Ambient |
MDL Number:
MFCD00077932 CAS číslo: 87-89-8 EINECS: 201-781-2 Merck Index: 14,4978 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...