Tisk…

myo-Inozitol 98+%

Dodavatel: Thermo Scientific

Synonyms: i-Inositol, D-Myo-Inositol, rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaol, Inositol

122261000 122265000 122260010 122260051
ACRO122261000EA 3320 CZK
ACRO122261000 ACRO122265000 ACRO122260010 ACRO122260051
myo-Inozitol 98+%
myo-Inozitol
Vzorec: C₆H₁₂O₆
MW: 180,16 g/mol
Bod varu: 291 °C (1013 hPa)
Bod tání: 220…228 °C
Hustota: 1,752 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00077932
CAS číslo: 87-89-8
EINECS: 201-781-2
Merck Index: 13,05001

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 220 to 228 °C
Loss on drying ≤0.5 %
Solubility (5 % in water) Clear colorless
HPLC ≥98.0 % (ELSD)
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR