Cyclohexyl isocyanate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
C105198-100G
C105198-25G
C105198-500G
SIALC105198-100GEA
1230
CZK
SIALC105198-100G
SIALC105198-25G
SIALC105198-500G
Cyclohexyl isocyanate, Sigma-Aldrich®
Cyclohexyl isocyanate
Vzorec:
C₇H₁₁NO Bod varu: 169 °C (1013 hPa) Bod tání: ∼–80 °C Hustota: 0,98 g/cm³ (20 °C) Bod vzplanutí: 53 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003840 CAS číslo: 3173-53-3 EINECS: 221-639-3 UN: 2488 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...