Tisk…

Cyclopropanecarbonyl chloride 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Cyclopropanecarboxylic chloride, Cyclopropanecarboxylic acid chloride

111630250 111631000 111630050 111635000
ACRO111630250EA 1720 CZK
ACRO111630250 ACRO111631000 ACRO111630050 ACRO111635000
Cyclopropanecarbonyl chloride 98%
Cyclopropanecarbonyl chloride
Vzorec: C₄H₅ClO
Bod varu: 119 °C (760 mmHg)
Hustota: 1,15 g/cm³
Bod vzplanutí: 23 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001277
CAS číslo: 4023-34-1
EINECS: 223-684-4
UN: 2920
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4515 to 1.4535 (20°C, 589 nm)
GC ≥97.5 %
Titration RCOCl ≥97.5 %
BSC Certification Passes
Microbial contamination (Salmonella) Absent in 375g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR