Cyclobutanecarbonyl chloride ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Cyclobutanecarboxylic acid chloride, Cyclobutanecarboxylic chloride, Cyclobutanecarbonylchloride
B20317.09
B20317.18
B20317.09EA
4330
CZK
B20317.09
B20317.18
Cyclobutanecarbonyl chloride ≥98%
Cyclobutanecarbonyl chloride
Purity: 98%
CAS No.: 5006-22-4
Molecular Formula: C5H7ClO
Formula Weight: 118.56
Flash Point: 37°(99°F)
Refractive Index: 1.4550
Odour: Stench
Boiling Point: 145°
Beilstein Reference: 956624
UN No.: 2920
MDL No.: MFCD00001319
Vzorec:
C₅H₇ClO MW: 118.56 Bod varu: 145 °C Hustota: 1.039 Bod vzplanutí: 37 °C (99 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001319 CAS číslo: 5006-22-4 EINECS: 225-680-8 UN: 2920 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...