Tisk…

Cyclobutanecarbonyl chloride 99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Cyclobutanecarboxylic acid chloride, Cyclobutanecarboxylic chloride, Cyclobutanecarbonylchloride

110910100 110910010 110910500
ACRO110910100EA 3830 CZK
ACRO110910100 ACRO110910010 ACRO110910500
Cyclobutanecarbonyl chloride 99%
Cyclobutanecarbonyl chloride
Vzorec: C₅H₇ClO
MW: 118.56
Bod varu: 145 °C (760 mmHg)
Bod vzplanutí: 37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001319
CAS číslo: 5006-22-4
EINECS: 225-680-8
UN: 2920
ADR: 8 II

Objednat

Výsledky specifikačního testu

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4545 to 1.4565 (20°C, 589 nm)
Titration RCOCl ≥98.5 %
Absorption Ratio P-15/P+15 1.06 to 1.19
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR