Tisk…

1,3-Cyclopentanedione, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Cyclopentane-1,3-dione

M-Clarity™ quality level = MQ100

177164-1G 177164-5G
SIAL177164-1GEA 2180 CZK
SIAL177164-1G SIAL177164-5G
1,3-Cyclopentanedione, Sigma-Aldrich®
1,3-Cyclopentanedione
Vzorec: C₅H₆O₂
MW: 98,1 g/mol
Bod tání: 149…152 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001405
CAS číslo: 3859-41-4
EINECS: 223-372-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR