Tisk…

Bromocyclopropane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Cyclopropyl bromide

M-Clarity™ quality level = MQ200

C117307-25G C117307-5G
SIALC117307-25GEA 11610 CZK
SIALC117307-25G SIALC117307-5G
Bromocyclopropane, Sigma-Aldrich®
Bromocyclopropane
Vzorec: C₃H₅Br
MW: 120,98 g/mol
Bod varu: 67…69 °C (1013 hPa)
Hustota: 1,505 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001271
CAS číslo: 4333-56-6
EINECS: 224-375-7
UN: 1993
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR