Bromocyclopentane, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Cyclopentyl bromide
MQ200
C115207-100G
C115207-250G
C115207-25G
SIALC115207-100GEA
1390
CZK
SIALC115207-100G
SIALC115207-250G
SIALC115207-25G
Bromocyclopentane, Sigma-Aldrich®
Bromocyclopentane
Vzorec:
C₅H₉Br MW: 149.03 Bod varu: 138 Hustota: 1.39 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001359 CAS číslo: 137-43-9 EINECS: 205-294-6 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...