Methyl-p-benzoquinone ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-Methylcyclohexa-2,5-diene-1,4-dione, Methylquinone, Methyl-1,4-benzoquinone
36670.09
36670.18
36670.09EA
1520
CZK
36670.09
36670.18
Methyl-p-benzoquinone ≥98%
Methyl-p-benzoquinone
Purity: 98%
CAS No.: 553-97-9
Molecular Formula: C7H6O2
Formula Weight: 122.12
Physical Form: Crystalline
Appearance: Yellowish green
Melting Point: 67-70°
MDL No.: MFCD00001603
Vzorec:
C₇H₆O₂ Bod varu: 187 °C (1013 hPa) Bod tání: 67…70 °C Hustota: 1,08 g/cm³ (75.5 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001603 CAS číslo: 553-97-9 EINECS: 209-056-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...