Tisk…

Kyanokobalamin, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

C3607-500MG C3607-5G
SIALC3607-500MGEA 10900 CZK
SIALC3607-500MG SIALC3607-5G
Kyanokobalamin, Sigma-Aldrich®
Kyanokobalamin
Vzorec: C₆₃H₈₈CoN₁₄O₁₄P
Bod tání: 320 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151092
CAS číslo: 68-19-9
EINECS: 200-680-0
Merck Index: 12,10152

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR