Kyanokobalamin ≥98% (dry weight)
Dodavatel: Thermo Fisher Scientific
A14894.03
A14894.06
A14894.14
A14894.03EA
3680
CZK
A14894.03
A14894.06
A14894.14
Kyanokobalamin ≥98% (dry weight)
Kyanokobalamin
Purity: 98+%
CAS No.: 68-19-9
Molecular Formula: C63H88CoN14O14P
Formula Weight: 1355.38
Storage Temperature: 2°C to 8°C
Merck Reference: 14,10014
MDL No.: MFCD00151092
Vzorec:
C₆₃H₈₈CoN₁₄O₁₄P Bod tání: 320 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00151092 CAS číslo: 68-19-9 EINECS: 200-680-0 Merck Index: 14,10014 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...