Tisk…

5-Methylquinoxaline, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ300

W320307-100G-K W320307-1KG-K W320307-250G-K
SIALW320307-100G-KEA 10460 CZK
SIALW320307-100G-K SIALW320307-1KG-K SIALW320307-250G-K
5-Methylquinoxaline, Sigma-Aldrich®
5-Methylquinoxaline
Vzorec: C₉H₈N₂
MW: 144,18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012335
CAS číslo: 13708-12-8
EINECS: 237-246-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR