4-Hydroxy-2-quinolinecarboxylic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Hydroxyquinoline-2-carboxylic Acid, Transtorine, Empirical Formula, Kynurenic acid, Quinurenic acid, KYNA, Kynuronic acid, Kinurenic acid
M-Clarity™ quality level = MQ200
K3375-1G
K3375-250MG
K3375-5G
SIALK3375-1GEA
2510
CZK
SIALK3375-1G
SIALK3375-250MG
SIALK3375-5G
4-Hydroxy-2-quinolinecarboxylic acid, Sigma-Aldrich®
4-Hydroxy-2-quinolinecarboxylic acid
Vzorec:
C₁₀H₇NO₃ MW: 189,17 g/mol Bod tání: 275 °C Hustota: 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149476 CAS číslo: 492-27-3 EINECS: 207-751-5 Merck Index: 13,05342 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...