Tisk…

3-Quinolinecarbaldehyde, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Quinoline-3-carboxaldehyde, Quinoline-3-carbaldehyde, 3-Quinolinecarboxaldehyde

M-Clarity™ quality level = MQ100

177121-1G 177121-5G
SIAL177121-1GEA 5480 CZK
SIAL177121-1G SIAL177121-5G
3-Quinolinecarbaldehyde, Sigma-Aldrich®
3-Quinolinecarbaldehyde
Vzorec: C₁₀H₇NO
MW: 157,17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00006768
CAS číslo: 13669-42-6
EINECS: 237-147-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR