2-Bromoquinoline 96%
Dodavatel: Thermo Scientific
|
Varování
|
H27302.03
H27302.06
H27302.14
H27302.03EA
2550
CZK
H27302.03
H27302.06
H27302.14
2-Bromoquinoline 96%
2-Bromoquinoline
Purity: 96%
CAS No.: 2005-43-8
Molecular Formula: C9H6BrN
Formula Weight: 208.06
MDL No.: MFCD00234480
Vzorec:
C₉H₆BrN MW: 208,06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00234480 CAS číslo: 2005-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...