(1S)-(-)-α-Pinene, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(-)-α-Pinene, (1S)-(-)-2-Pinene, (-)-Pin-2(3)-ene, L(-)-α-Pinene
M-Clarity™ quality level = MQ100
P45702-100ML
P45702-500ML
SIALP45702-100MLEA
915
CZK
SIALP45702-100ML
SIALP45702-500ML
(1S)-(-)-α-Pinene, Sigma-Aldrich®
(1S)-(-)-α-Pinene
Vzorec:
C₁₀H₁₆ MW: 136.24 Bod varu: 155 Bod tání: –62 °C Hustota: 0.8592 g/cm³ (20 °C) Bod vzplanutí: 32 Storage Temperature: Refrigerator |
MDL Number:
MFCD00064145 CAS číslo: 7785-26-4 EINECS: 232-077-3 UN: 2368 ADR: 3 III Merck Index: 12,07599 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...