Tisk…

(+)-3-Carene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

441619-1G 441619-5G
SIAL441619-1GEA 2980 CZK
SIAL441619-1G SIAL441619-5G
(+)-3-Carene, Sigma-Aldrich®
(+)-3-Carene
Vzorec: C₁₀H₁₆
Storage Temperature: Ambient
MDL Number: MFCD00066417
CAS číslo: 498-15-7
EINECS: 207-856-6
UN: 2319
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR