Tisk…

5-Methylbenzotriazole, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5-Methyl-1H-benzotriazole

M-Clarity™ quality level = MQ200

196304-10G 196304-50G
SIAL196304-10GEA 1330 CZK
SIAL196304-10G SIAL196304-50G
5-Methylbenzotriazole, Sigma-Aldrich®
5-Methylbenzotriazole
Vzorec: C₇H₇N₃
MW: 133,15 g/mol
Bod varu: 361 °C (1013 hPa)
Bod tání: 85 °C
MDL Number: MFCD00005702
CAS číslo: 136-85-6
EINECS: 205-265-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR