Tisk…

5-Methylbenzotriazole ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 5-Methyl-1H-benzotriazole

A10615.22 A10615.36
A10615.22EA 4670 CZK
A10615.22 A10615.36
5-Methylbenzotriazole ≥98%
5-Methylbenzotriazole

Purity: 98+% CAS No.: 136-85-6 Molecular Formula: C7H7N3 Formula Weight: 133.15 Odour: Odourless pH: 5.3 Melting Point: 77-84° Boiling Point: 210-212°/12mm Beilstein Reference: 116658 MDL No.: MFCD00005702

Vzorec: C₇H₇N₃
MW: 133,15 g/mol
Bod varu: 210…212 °C (12 mmHg)
Bod tání: 77…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00005702
CAS číslo: 136-85-6
EINECS: 205-265-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR