Tisk…

Benzo[e]pyrene, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: BeP

M-Clarity™ quality level = MQ100

B10102-100MG B10102-25MG
SIALB10102-100MGEA 14730 CZK
SIALB10102-100MG SIALB10102-25MG
Benzo[e]pyrene, Sigma-Aldrich®
Benzo[e]pyrene
Vzorec: C₂₀H₁₂
MW: 252,32 g/mol
Bod varu: 492 °C (1013 hPa)
Bod tání: 178…179 °C
CAS číslo: 192-97-2
EINECS: 205-892-7
UN: 3077
ADR: 9,III
Merck Index: 12,01135

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR