Tisk…

6-Methylcoumarin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 6-Methyl-2-benzopyrone, 6-Methyl-2H-chromen-2-one

M-Clarity™ quality level = MQ300

W269905-100G-K W269905-1KG-K W269905-5KG-K
SIALW269905-100G-KEA 0 CZK
SIALW269905-100G-K SIALW269905-1KG-K SIALW269905-5KG-K
6-Methylcoumarin, Sigma-Aldrich®
6-Methylcoumarin
Vzorec: C₁₀H₈O₂
MW: 160,17 g/mol
Bod varu: 303 °C (1013 hPa)
Bod tání: 72…74 °C
MDL Number: MFCD00006875
CAS číslo: 92-48-8
EINECS: 202-158-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR