6-Methylcoumarin ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
6-Methyl-2-benzopyrone, 6-Methyl-2H-chromen-2-one
A19382.18
A19382.22
A19382.18EA
1300
CZK
A19382.18
A19382.22
6-Methylcoumarin ≥99%
6-Methylcoumarin
Purity: 99%
CAS No.: 92-48-8
Molecular Formula: C10H8O2
Formula Weight: 160.17
Melting Point: 74-77°
Boiling Point: 303°/725mm
MDL No.: MFCD00006875
Vzorec:
C₁₀H₈O₂ MW: 160,17 g/mol Bod varu: 303 °C (725 mmHg) Bod tání: 74…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006875 CAS číslo: 92-48-8 EINECS: 202-158-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...