Tisk…

4-Hydroxycoumarin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Hydroxy-2-benzopyrone

M-Clarity™ quality level = MQ100

H23805-100G H23805-500G
SIALH23805-100GEA 1670 CZK
SIALH23805-100G SIALH23805-500G
4-Hydroxycoumarin, Sigma-Aldrich®
4-Hydroxycoumarin
Vzorec: C₉H₆O₃
MW: 162,14 g/mol
Bod varu: 349 °C (1013 hPa)
Bod tání: 211…214 °C
MDL Number: MFCD00006856
CAS číslo: 1076-38-6
EINECS: 214-060-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR