Tisk…

Kumarin ≥98%, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ300

W526509-10KG W526509-SAMPLE
SIALW526509-10KGEA 0 CZK
SIALW526509-10KG SIALW526509-SAMPLE
Kumarin ≥98%, Sigma-Aldrich®
Kumarin

Coumarin is an aroma compound responsible for the characteristic odor of hay. It is one of the major volatile compounds found in the aerial parts of bastard balm.


  • biological source: synthetic
  • grade: Halal

Vzorec: C₉H₆O₂
Bod varu: 298 °C (lit.)
Bod tání: 67…73 °C (lit.)
Hustota: 0,935 g/cm³ (20 °C)
Bod vzplanutí: 162 °C
Storage Temperature: Ambient
MDL Number: MFCD00006850
CAS číslo: 91-64-5
EINECS: 202-086-7
UN: 2811
ADR: 6.1,III
Merck Index: 13,02588

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR