Kumarin ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A15336.0B
A15336.0I
A15336.30
A15336.0BEA
4600
CZK
A15336.0B
A15336.0I
A15336.30
Kumarin ≥98%
Kumarin
Purity: 98%
CAS No.: 91-64-5
Molecular Formula: C9H6O2
Formula Weight: 146.15
Flash Point: 162°(323°F)
Odour: Pleasant
Boiling Point: 297-299°
Beilstein Reference: 383644
Merck Reference: 14,2562
UN No.: 2811
MDL No.: MFCD00006850
Vzorec:
C₉H₆O₂ Bod varu: 297…299 °C Bod tání: 68…71 °C Hustota: 0,935 Bod vzplanutí: 162 °C (323 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006850 CAS číslo: 91-64-5 EINECS: 202-086-7 UN: 2811 ADR: 6.1,III Merck Index: 14,2562 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...