Tisk…

p-Terphenyl, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Phenyl-1,1'-biphenyl, 1,4-Diphenylbenzene, 4-Phenylbiphenyl, 1,4-Terphenyl

M-Clarity™ quality level = MQ100

T3203-100G T3203-25G
SIALT3203-100GEA 6300 CZK
SIALT3203-100G SIALT3203-25G
p-Terphenyl, Sigma-Aldrich®
p-Terphenyl
Vzorec: C₁₈H₁₄
MW: 230,31 g/mol
Bod varu: 383 °C (1013 hPa)
Bod tání: 210…211 °C
Hustota: 1,234 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003061
CAS číslo: 92-94-4
EINECS: 202-205-2
UN: 3077
ADR: 9,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR