p-Terphenyl ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-Phenyl-1,1'-biphenyl, 1,4-Diphenylbenzene, 4-Phenylbiphenyl, 1,4-Terphenyl
A14833.14
A14833.22
A14833.30
A14833.14EA
1240
CZK
A14833.14
A14833.22
A14833.30
p-Terphenyl ≥99%
p-Terphenyl
Purity: 99+%
CAS No.: 92-94-4
Molecular Formula: C18H14
Formula Weight: 230.31
Flash Point: 207°(404°F)
Melting Point: 212-215°
Boiling Point: 405°
Beilstein Reference: 1908447
MDL No.: MFCD00003061
Vzorec:
C₁₈H₁₄ MW: 230,31 g/mol Bod varu: 405 °C Bod tání: 212…215 °C Hustota: 1,23 Bod vzplanutí: 207 °C (404 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003061 CAS číslo: 92-94-4 EINECS: 202-205-2 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...