Bifenyl-4-ol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
[1,1'-Biphenyl]-4-ol, 4-Hydroxydiphenyl, 4-Phenylphenol, p-Hydroxybiphenyl, 4-Hydroxybiphenyl
M-Clarity™ quality level = MQ100
134341-100G
134341-5G
SIAL134341-100GEA
833
CZK
SIAL134341-100G
SIAL134341-5G
Bifenyl-4-ol, Sigma-Aldrich®
Bifenyl-4-ol
Vzorec:
C₁₂H₁₀O MW: 170.21 Bod varu: 321 Bod tání: 162 Hustota: 1.29 g/cm³ (20 °C) |
MDL Number:
MFCD00002347 CAS číslo: 92-69-3 EINECS: 202-179-2 Merck Index: 12,07459 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...