2-Fluorobiphenyl, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Fluoro-1,1'-biphenyl, PFB No. 1
M-Clarity™ quality level = MQ100
102741-10G
102741-1G
SIAL102741-10GEA
4590
CZK
SIAL102741-10G
SIAL102741-1G
2-Fluorobiphenyl, Sigma-Aldrich®
2-Fluorobiphenyl
Vzorec:
C₁₂H₉F MW: 172.2 Bod varu: 248 Bod tání: 73…74 °C Hustota: 1.2452 g/cm³ (25 °C) |
MDL Number:
MFCD00000317 CAS číslo: 321-60-8 EINECS: 206-290-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...