Tisk…

Benzyl salicylate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

84260-100ML 84260-500ML
SIAL84260-100MLEA 1220 CZK
SIAL84260-100ML SIAL84260-500ML
Benzyl salicylate, Sigma-Aldrich®
Benzyl salicylate
Vzorec: C₁₄H₁₂O₃
Bod varu: 208 °C (1013 hPa)
Hustota: 1,1755 g/cm³ (20 °C)
MDL Number: MFCD00020034
CAS číslo: 118-58-1
EINECS: 204-262-9
Merck Index: 13,01146

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR