4-Phenylbenzonitrile 95%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
4-Biphenylcarbonitrile, 4-Cyanobiphenyl, p-Phenylbenzonitrile, [1,1'-Biphenyl]-4-carbonitrile
H26791.09
H26791.18
H26791.09EA
3640
CZK
H26791.09
H26791.18
4-Phenylbenzonitrile 95%
4-Phenylbenzonitrile
Purity: 95%
CAS No.: 2920-38-9
Molecular Formula: C13H9N
Formula Weight: 179.22
Flash Point:
Vzorec:
C₁₃H₉N MW: 179,22 g/mol Bod tání: 85…87 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001821 CAS číslo: 2920-38-9 EINECS: 220-860-2 UN: 3439 ADR: 6.1,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...