4-(Benzyloxy)benzaldehyde, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
123714-100G
123714-25G
SIAL123714-100GEA
2420
CZK
SIAL123714-100G
SIAL123714-25G
4-(Benzyloxy)benzaldehyde, Sigma-Aldrich®
4-(Benzyloxy)benzaldehyde
Vzorec:
C₁₄H₁₂O₂ MW: 212,25 g/mol Bod varu: 366 °C (1013 hPa) Bod tání: 70…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003387 CAS číslo: 4397-53-9 EINECS: 224-527-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...