4-Bromobiphenyl (PBB No. 3) ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
4-Bromodiphenyl, PBB No. 3, 4-Bromobiphenyl
A12638.09
A12638.18
A12638.30
A12638.09EA
1320
CZK
A12638.09
A12638.18
A12638.30
4-Bromobiphenyl (PBB No. 3) ≥98%
4-Bromobiphenyl (PBB No. 3)
Purity: 98+%
CAS No.: 92-66-0
Molecular Formula: C12H9Br
Formula Weight: 233.12
Flash Point: 143°(289°F)
Melting Point: 88-92°
Boiling Point: 310°
Beilstein Reference: 1907453
MDL No.: MFCD00000100
Vzorec:
C₁₂H₉Br Bod varu: 310 °C Bod tání: 88…92 °C Hustota: 1,615 g/cm³ (20 °C) Bod vzplanutí: 143 °C (289 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000100 CAS číslo: 92-66-0 EINECS: 202-176-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...