Tisk…

4,4'-Dibromobiphenyl (PBB No. 15), Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: PBB No. 15

M-Clarity™ quality level = MQ100

229237-100G 229237-25G
SIAL229237-100GEA 2450 CZK
SIAL229237-100G SIAL229237-25G
4,4'-Dibromobiphenyl (PBB No. 15), Sigma-Aldrich®
4,4'-Dibromobiphenyl (PBB No. 15)
Vzorec: C₁₂H₈Br₂
Bod varu: 357 °C (1013 hPa)
Bod tání: 164 °C
Hustota: 1,867 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000101
CAS číslo: 92-86-4
EINECS: 202-198-6
UN: 3152
ADR: 9,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR