Tisk…

4,4'-Dibromobiphenyl (PBB No. 15) ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: PBB No. 15

A14139.18 A14139.30
A14139.18EA 1430 CZK
A14139.18 A14139.30
4,4'-Dibromobiphenyl (PBB No. 15) ≥98%
4,4'-Dibromobiphenyl (PBB No. 15)

Purity: 98+% CAS No.: 92-86-4 Molecular Formula: C12H8Br2 Formula Weight: 312.02 Melting Point: 164-169° Boiling Point: 355-360° UN No.: 3152 MDL No.: MFCD00000101

Vzorec: C₁₂H₈Br₂
Bod varu: 355…360 °C
Bod tání: 164…169 °C
Hustota: 1,867 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000101
CAS číslo: 92-86-4
EINECS: 202-198-6
UN: 3152
ADR: 9,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR