2,4,6-Trimethylbenzylamine 99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Mesitylmethanamine, 1-Mesitylmethanamine
351180010
351180050
ACRO351180010EA
1240
CZK
ACRO351180010
ACRO351180050
2,4,6-Trimethylbenzylamine 99%
2,4,6-Trimethylbenzylamine
Vzorec:
C₁₀H₁₅N MW: 149.24 |
MDL Number:
MFCD03411013 CAS číslo: 40393-99-5 UN: 2735 |
Výsledky specifikačního testu
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5360 to 1.5380 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...