Tisk…

2,4,6-Triisopropylbenzenesulphonyl chloride, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: 2,4,6-Triisopropylbenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ200

119490-100G 119490-25G
SIAL119490-100GEA 6430 CZK
SIAL119490-100G SIAL119490-25G
2,4,6-Triisopropylbenzenesulphonyl chloride, Sigma-Aldrich®
2,4,6-Triisopropylbenzenesulphonyl chloride
Vzorec: C₁₅H₂₃ClO₂S
MW: 302,87 g/mol
Bod tání: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00007433
CAS číslo: 6553-96-4
EINECS: 229-479-6
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR